Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005729.3 | NP_005720.1 | 2 | 293 | ||
XM_005269379.2 | XP_005269436.1 | 2 | 291 | |||
GenBank mRNA | AK296669.1 | - | 2 | 286 | ||
BC005020.1 | - | 2 | 279 | 3659599 | ||
BC013204.1 | - | 2 | 270 | |||
BC110299.1 | - | 2 | 270 | 5215228 | ||
BE791220.1 | - | 2 | 309 | |||
M80254.1 | - | 2 | 291 |