Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF20A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005733.2 | NP_005724.1 | 8 | 1434 | ||
GenBank mRNA | AF070672.1 | - | 7 | 965 | ||
AF153329.1 | - | 8 | 972 | |||
AK025790.1 | - | 8 | 993 | |||
AK296879.1 | - | 9 | 948 | |||
BC012999.2 | - | 8 | 972 | 3604291 |