Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031889.2 | NP_114095.2 | 9 | 1563 | ||
XM_006714056.3 | XP_006714119.1 | 8 | 3312 | |||
GenBank mRNA | AF125373.1 | - | 9 | 1563 | ||
BC117308.1 | - | 8 | 1352 | 40125859 | ||
BC117310.1 | - | 8 | 1352 | 40125861 |