Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005736.3 | NP_005727.1 | 4 | 349 | ||
GenBank mRNA | AK297362.1 | - | 4 | 312 | ||
AK297620.1 | - | 3 | 243 | |||
AK302072.1 | - | 3 | 238 | |||
AK312506.1 | - | 4 | 308 | |||
BC000693.2 | - | 4 | 285 | 3347881 | ||
BC026016.2 | - | 4 | 251 | 3009822 | ||
X82206.1 | - | 4 | 307 | |||
Z14978.1 | - | 4 | 289 |