Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NAMPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005746.2 | NP_005737.1 | 1, 2 | 366 | ||
XM_005250100.2 | XP_005250157.1 | 2, 3 | 347 | |||
GenBank mRNA | AK292851.1 | - | 1, 2 | 126 | ||
BC072439.1 | - | 1, 2 | 139 | 6023159 | ||
BC106046.1 | - | 1, 2 | 162 | 6161081 | ||
U02020.1 | - | 1, 2 | 85 |