Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT1HL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276687.1 | NP_001263616.1 | 1 | 267 | ||
GenBank mRNA | AF333388.1 | - | 1 | 267 |
Gene Symbol: MT1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005951.2 | NP_005942.1 | 3 | 292 | ||
GenBank mRNA | BC008408.1 | - | 3 | 292 | 4274128 | |
BC059780.1 | - | 3 | 272 | 5227592 | ||
X64177.1 | - | 3 | 244 |