Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZK1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005764.3 | NP_005755.1 | 3, 4 | 447 | ||
GenBank mRNA | BC012303.1 | - | 3, 4 | 360 | 4469903 | |
BI763294.1 | - | 1, 2 | 112 | |||
BM924665.1 | - | 3, 4 | 473 | |||
CR450304.1 | - | 3, 4 | 280 |