Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM72C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287385.1 | NP_001274314.1 | 1 | 993 | ||
XM_011509724.2 | XP_011508026.1 | 1 | 993 | |||
XM_017001682.1 | XP_016857171.1 | 2 | 1243 |
Gene Symbol: FAM72A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123168.2 | NP_001116640.1 | 1 | 978 | ||
XM_006711526.3 | XP_006711589.1 | 2 | 952 | |||
XM_011509966.2 | XP_011508268.1 | 1 | 993 | |||
XM_017002233.1 | XP_016857722.1 | 2 | 858 | |||
GenBank mRNA | BC035696.1 | - | 1 | 545 | 5541342 | |
BC137545.1 | - | 1 | 509 | 9021545 | ||
BC146978.1 | - | 1 | 977 | 9056906 | ||
BC146992.1 | - | 1 | 976 | 9056920 |
Gene Symbol: FAM72B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100910.1 | NP_001094380.1 | 1 | 957 | ||
GenBank mRNA | BC137546.1 | - | 1 | 510 | 9021546 |
Gene Symbol: FAM72D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207418.2 | NP_997301.2 | 1 | 976 | ||
XM_011509963.2 | XP_011508265.1 | 1 | 993 | |||
XM_017002227.1 | XP_016857716.1 | 3 | 1396 | |||
GenBank mRNA | AB096683.1 | - | 1 | 976 | ||
AK307197.1 | - | 1 | 952 |