Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033103.4 | NP_149094.3 | 11 | 1378 | ||
GenBank mRNA | AF423421.1 | - | 11 | 1312 | ||
AJ347750.1 | - | 11 | 1312 | |||
AK095001.1 | - | 2 | 148 | |||
AK098246.1 | - | 11 | 1348 | |||
AK126506.1 | - | 8 | 1646 | |||
AK300775.1 | - | 10 | 1272 | |||
AK315105.1 | - | 11 | 1348 | |||
AL831950.1 | - | 10 | 1226 | |||
BC036447.2 | - | 11 | 1348 | 4830913 |