Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005264783.4 | XP_005264840.1 | 2 | 343 | ||
XM_017005495.1 | XP_016860984.1 | 2 | 315 | |||
GenBank mRNA | AF011573.1 | - | 1 | 130 | ||
AK314196.1 | - | 2 | 311 | |||
AY074878.1 | - | 2 | 311 | |||
AY261677.1 | - | 1 | 144 | |||
AY279353.1 | - | 2 | 332 | |||
BC017584.1 | - | 1 | 296 | |||
Z21707.1 | - | 1 | 45 |