Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ULK4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026858.1 | 5 | 322 | |||
GenBank mRNA | BC071630.1 | - | 5 | 322 | ||
BC137210.1 | - | 6 | 416 |
Gene Symbol: ULK4P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027470.1 | 5 | 322 | |||
GenBank mRNA | BC137222.1 | - | 4 | 305 |
Gene Symbol: ULK4P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026859.1 | 5 | 581 | |||
GenBank mRNA | BC023564.2 | - | 5 | 581 | ||
BC071855.1 | - | 5 | 579 |