Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC441155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271675.1 | NP_001258604.1 | 1, 2 | 228 | ||
GenBank mRNA | AK302514.1 | - | 1, 2 | 185 | ||
DB093153.1 | - | 1, 2 | 230 |