Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163278.1 | NP_001156750.1 | 30 | 6333 | ||
NM_001163279.1 | NP_001156751.1 | 30 | 6330 | |||
NM_014253.3 | NP_055068.2 | 29 | 6312 | |||
XM_011531230.2 | XP_011529532.1 | 33 | 6974 | |||
XM_011531236.2 | XP_011529538.2 | 19 | 5551 | |||
XM_011531237.2 | XP_011529539.1 | 9 | 2435 | |||
XM_017029208.1 | XP_016884697.1 | 34 | 7073 | |||
XM_017029209.1 | XP_016884698.1 | 35 | 7128 | |||
XM_017029210.1 | XP_016884699.1 | 33 | 7001 | |||
XM_017029211.1 | XP_016884700.1 | 34 | 7079 | |||
XM_017029212.1 | XP_016884701.1 | 34 | 6690 | |||
XM_017029213.1 | XP_016884702.1 | 33 | 7002 | |||
XM_017029214.1 | XP_016884703.1 | 32 | 6953 | |||
GenBank mRNA | AF100772.1 | - | 29 | 6312 | ||
BC140783.1 | - | 30 | 6330 | 9021651 | ||
BC144543.1 | - | 30 | 6327 | 9053070 |