Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD36B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005263995.4 | XP_005264052.1 | 1 | 193 | ||
XM_006712661.1 | XP_006712724.1 | 1 | 193 | |||
XM_011511555.1 | XP_011509857.1 | 1 | 193 | |||
XM_011511556.1 | XP_011509858.1 | 1 | 193 | |||
XM_011511557.2 | XP_011509859.1 | 1 | 193 | |||
XM_011511558.1 | XP_011509860.1 | 1 | 193 | |||
XM_017004578.1 | XP_016860067.1 | 1 | 193 | |||
XM_017004579.1 | XP_016860068.1 | 1 | 193 | |||
XM_017004580.1 | XP_016860069.1 | 1 | 193 | |||
XM_017004581.1 | XP_016860070.1 | 1 | 193 | |||
XM_017004582.1 | XP_016860071.1 | 1 | 193 | |||
XM_017004583.1 | XP_016860072.1 | 1 | 193 | |||
XM_017004584.1 | XP_016860073.1 | 1 | 193 | |||
XM_017004585.1 | XP_016860074.1 | 1 | 193 | |||
XM_017004587.1 | XP_016860076.1 | 1 | 193 | |||
XM_017004588.1 | XP_016860077.1 | 1 | 193 | |||
XM_017004589.1 | XP_016860078.1 | 1 | 193 | |||
XM_017004590.1 | XP_016860079.1 | 1 | 193 | |||
XM_017004592.1 | XP_016860081.1 | 1 | 193 | |||
XM_017004593.1 | XP_016860082.1 | 1 | 193 | |||
XM_017004594.1 | XP_016860083.1 | 1 | 193 | |||
XM_017004595.1 | XP_016860084.1 | 1 | 193 |
Gene Symbol: ANKRD36C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310154.1 | NP_001297083.1 | 1 | 281 |