Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALG3 SIRNA Assays ›
Gene Symbol:
ALG3
Gene Name:
ALG3, alpha-1,3- mannosyltransferase
Gene Aliases:
CDG1D, CDGS4, CDGS6, D16Ertd36e, NOT56L, Not56, not
Chromosome Location:
Chr.3: 184242329 - 184249525 on Build GRCh38
UniGene ID:
Hs.478481
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s19866
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ALG3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001006941.2
NP_001006942.1
3
416
NM_005787.5
NP_005778.1
3
343
NR_024533.1
2
243
NR_024534.1
3
306
XM_011512322.1
XP_011510624.1
3
228
XM_011512323.2
XP_011510625.1
3
214
GenBank mRNA
AK289361.1
-
3
372
AK297701.1
-
2
197
AK299575.1
-
2
234
BC002839.2
-
3
318
3636604
BC004313.1
-
3
298
BC021062.1
-
3
307
BM920198.1
-
3
272
BX339649.2
-
3
421
KP189279.1
-
3
336
KP189280.1
-
3
299
KP189281.1
-
2
152
KP189282.1
-
3
453
KP189283.1
-
2
236
KP189284.1
-
3
334
KP189285.1
-
3
297
KP189286.1
-
2
152
KP189287.1
-
3
252
KP189288.1
-
2
234
KP189289.1
-
3
333
KP189290.1
-
3
297
KP189291.1
-
2
152
KP189292.1
-
3
252
KP189293.1
-
2
234
KP189294.1
-
3
312
KP189295.1
-
3
312
KP189296.1
-
3
275
KP189297.1
-
3
275
KP189298.1
-
1
26
KP189299.1
-
3
168
KP189300.1
-
2
212
KP189301.1
-
3
275
KP189302.1
-
3
333
KP189303.1
-
3
295
KP189304.1
-
2
152
KP189305.1
-
3
252
KP189306.1
-
2
232
KP189307.1
-
3
332
KP189308.1
-
3
295
KP189309.1
-
2
152
KP189310.1
-
3
252
KP189311.1
-
2
232
KP189312.1
-
3
332
KP189313.1
-
3
332
KP189314.1
-
3
332
KP189315.1
-
3
295
KP189316.1
-
3
295
KP189317.1
-
3
295
KP189318.1
-
2
152
KP189319.1
-
2
152
KP189320.1
-
2
152
KP189321.1
-
3
252
KP189322.1
-
3
252
KP189323.1
-
3
252
KP189324.1
-
2
232
KP189325.1
-
2
232
KP189326.1
-
2
232
KP189327.1
-
2
232
Y09022.1
-
3
343
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00271780_m1
Panther Classification:
Molecular Function -
glycosyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
protein glycosylation
dolichol-linked oligosaccharide biosynthetic process
mannosylation
alpha-1,3-mannosyltransferase activity
dol-P-Man:Man(5)GlcNAc(2)-PP-Dol alpha-1,3-mannosyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0