Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006941.2 | NP_001006942.1 | 7, 8 | 1122 | ||
NM_005787.5 | NP_005778.1 | 7, 8 | 1049 | |||
NR_024533.1 | 6, 7 | 949 | ||||
NR_024534.1 | 7, 8 | 1012 | ||||
XM_011512322.1 | XP_011510624.1 | 7, 8 | 934 | |||
XM_011512323.2 | XP_011510625.1 | 7, 8 | 920 | |||
GenBank mRNA | AK289361.1 | - | 7, 8 | 1078 | ||
AK299575.1 | - | 6, 7 | 940 | |||
BC002839.2 | - | 7, 8 | 1024 | 3636604 | ||
BC004313.1 | - | 7, 8 | 1004 | |||
BC021062.1 | - | 7, 8 | 1013 | |||
KP189279.1 | - | 7, 8 | 1042 | |||
KP189280.1 | - | 7, 8 | 1005 | |||
KP189281.1 | - | 6, 7 | 858 | |||
KP189282.1 | - | 7, 8 | 1159 | |||
KP189283.1 | - | 6, 7 | 942 | |||
KP189284.1 | - | 7, 8 | 1040 | |||
KP189285.1 | - | 7, 8 | 1003 | |||
KP189286.1 | - | 6, 7 | 858 | |||
KP189287.1 | - | 7, 8 | 958 | |||
KP189288.1 | - | 6, 7 | 940 | |||
KP189289.1 | - | 7, 8 | 1039 | |||
KP189290.1 | - | 7, 8 | 1003 | |||
KP189291.1 | - | 6, 7 | 858 | |||
KP189292.1 | - | 7, 8 | 958 | |||
KP189293.1 | - | 6, 7 | 940 | |||
KP189294.1 | - | 7, 8 | 1018 | |||
KP189295.1 | - | 6, 7 | 812 | |||
KP189296.1 | - | 7, 8 | 981 | |||
KP189297.1 | - | 6, 7 | 775 | |||
KP189298.1 | - | 4, 5 | 571 | |||
KP189299.1 | - | 7, 8 | 874 | |||
KP189300.1 | - | 6, 7 | 918 | |||
KP189302.1 | - | 7, 8 | 1039 | |||
KP189303.1 | - | 7, 8 | 1001 | |||
KP189304.1 | - | 6, 7 | 858 | |||
KP189305.1 | - | 7, 8 | 958 | |||
KP189306.1 | - | 6, 7 | 938 | |||
KP189307.1 | - | 7, 8 | 1038 | |||
KP189308.1 | - | 7, 8 | 1001 | |||
KP189309.1 | - | 6, 7 | 858 | |||
KP189310.1 | - | 7, 8 | 958 | |||
KP189311.1 | - | 6, 7 | 938 | |||
KP189312.1 | - | 7, 8 | 1038 | |||
KP189313.1 | - | 6, 7 | 832 | |||
KP189315.1 | - | 7, 8 | 1001 | |||
KP189316.1 | - | 6, 7 | 795 | |||
KP189318.1 | - | 6, 7 | 858 | |||
KP189319.1 | - | 5, 6 | 652 | |||
KP189321.1 | - | 7, 8 | 958 | |||
KP189322.1 | - | 6, 7 | 752 | |||
KP189324.1 | - | 6, 7 | 938 | |||
KP189325.1 | - | 5, 6 | 732 | |||
Y09022.1 | - | 7, 8 | 1049 |