Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267045.1 | NP_001253974.1 | 6 | 968 | ||
NM_005789.3 | NP_005780.2 | 4 | 641 | |||
NM_176863.2 | NP_789839.1 | 4 | 641 | |||
NR_049772.1 | 4 | 641 | ||||
XM_011524177.2 | XP_011522479.1 | 4 | 650 | |||
GenBank mRNA | AK057221.1 | - | 6 | 807 | ||
AK074999.1 | - | 4 | 338 | |||
AK292618.1 | - | 4 | 366 | |||
AK303211.1 | - | 4 | 338 | |||
AK307110.1 | - | 3 | 1042 | |||
AK314347.1 | - | 4 | 384 | |||
BC001423.2 | - | 4 | 368 | 3139753 | ||
BC002684.1 | - | 4 | 347 | 3606558 | ||
BC008020.1 | - | 4 | 299 | 3537911 | ||
BG708969.1 | - | 4 | 382 | |||
BI464061.1 | - | 4 | 644 | |||
BT019386.1 | - | 4 | 151 | |||
BX640801.1 | - | 6 | 695 | |||
CR541745.1 | - | 4 | 151 | |||
EU794653.1 | - | 4 | 368 | |||
FJ200488.1 | - | 4 | 151 | |||
JX156303.1 | - | 4 | 151 | |||
U11292.1 | - | 4 | 313 |