Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267045.1 | NP_001253974.1 | 9 | 1259 | ||
NM_005789.3 | NP_005780.2 | 7 | 932 | |||
NM_176863.2 | NP_789839.1 | 7 | 971 | |||
NR_049772.1 | 6 | 819 | ||||
XM_011524177.2 | XP_011522479.1 | 7 | 941 | |||
GenBank mRNA | AK057221.1 | - | 9 | 1098 | ||
AK074999.1 | - | 7 | 629 | |||
AK292618.1 | - | 7 | 657 | |||
AK303211.1 | - | 6 | 516 | |||
AK307110.1 | - | 6 | 1333 | |||
AK314347.1 | - | 7 | 675 | |||
BC001423.2 | - | 7 | 659 | 3139753 | ||
BC002684.1 | - | 7 | 638 | 3606558 | ||
BC008020.1 | - | 7 | 590 | 3537911 | ||
BG708969.1 | - | 7 | 673 | |||
BT019386.1 | - | 7 | 442 | |||
BX640801.1 | - | 9 | 987 | |||
CR541745.1 | - | 7 | 442 | |||
EU794653.1 | - | 7 | 659 | |||
JX156303.1 | - | 5 | 190 | |||
U11292.1 | - | 7 | 643 |