Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308426.1 | NP_001295355.1 | 5 | 698 | ||
NM_001308427.1 | NP_001295356.1 | 5 | 507 | |||
NM_001308428.1 | NP_001295357.1 | 5 | 498 | |||
NM_001308430.1 | NP_001295359.1 | 4 | 441 | |||
NM_001308431.1 | NP_001295360.1 | 4 | 658 | |||
NM_001308433.1 | NP_001295362.1 | 4 | 458 | |||
NM_001308434.1 | NP_001295363.1 | 4 | 601 | |||
NM_005793.4 | NP_005784.1 | 5 | 481 | |||
XM_017005511.1 | XP_016861000.1 | 4 | 369 | |||
XM_017005512.1 | XP_016861001.1 | 2 | 1032 | |||
XM_017005513.1 | XP_016861002.1 | 5 | 423 | |||
XM_017005514.1 | XP_016861003.1 | 6 | 680 | |||
XM_017005515.1 | XP_016861004.1 | 4 | 436 | |||
XM_017005516.1 | XP_016861005.1 | 4 | 398 | |||
XM_017005517.1 | XP_016861006.1 | 2 | 198 | |||
XM_017005518.1 | XP_016861007.1 | 3 | 327 | |||
GenBank mRNA | AF051941.1 | - | 5 | 414 | ||
AK222555.1 | - | 4 | 380 | |||
AK294809.1 | - | 4 | 414 | |||
AK308970.1 | - | 2 | 185 | |||
BC001808.2 | - | 5 | 428 | 2966629 | ||
BC012828.2 | - | 5 | 428 | 2966629 | ||
BE866839.1 | - | 5 | 449 | |||
BG284082.1 | - | 4 | 387 | |||
BG530435.1 | - | 4 | 303 | |||
CT002144.1 | - | 4 | 658 | |||
DA398008.1 | - | 5 | 454 | |||
U90449.1 | - | 5 | 428 |