Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258038.1 | NP_001244967.1 | 1, 2 | 134 | ||
XM_005262686.1 | XP_005262743.1 | 1, 2 | 138 | |||
GenBank mRNA | AL602431.1 | - | 1, 2 | 135 | ||
DA765300.1 | - | 1, 2 | 105 |