Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005805.5 | NP_005796.1 | 5 | 617 | ||
GenBank mRNA | AK055128.1 | - | 5 | 854 | ||
BC066336.1 | - | 5 | 463 | 5296432 | ||
U86782.1 | - | 5 | 349 |