Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RASGRP2 SIRNA Assays ›
Gene Symbol:
RASGRP2
Gene Name:
RAS guanyl releasing protein 2
Gene Aliases:
CALDAG-GEFI, CDC25L
Chromosome Location:
Chr.11: 64726911 - 64745456 on Build GRCh38
UniGene ID:
Hs.99491
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s19977
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RASGRP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098670.1
NP_001092140.1
9, 10
1289
NM_001098671.1
NP_001092141.1
9, 10
1329
NM_001318398.1
NP_001305327.1
9, 10
1249
NM_153819.1
NP_722541.1
9, 10
1355
XM_005273707.3
XP_005273764.3
9, 10
1680
XM_011544718.1
XP_011543020.1
9, 10
1680
XM_011544720.1
XP_011543022.1
9, 10
1760
XM_011544721.1
XP_011543023.1
9, 10
1292
XM_011544722.1
XP_011543024.1
9, 10
1345
XM_011544723.2
XP_011543025.1
9, 10
1294
XM_011544725.2
XP_011543027.1
8, 9
1126
XM_017017082.1
XP_016872571.1
8, 9
2330
XM_017017083.1
XP_016872572.1
8, 9
2330
XM_017017084.1
XP_016872573.1
9, 10
1323
XM_017017085.1
XP_016872574.1
6, 7
1113
XM_017017086.1
XP_016872575.1
6, 7
829
GenBank mRNA
AF043722.1
-
10, 11
1461
AF043723.1
-
9, 10
1355
AF081194.1
-
9, 10
1262
AK092852.1
-
4, 5
1451
AK092882.1
-
8, 9
2025
AK122694.1
-
9, 10
1213
AK314637.1
-
8, 9
1240
BC110306.1
-
9, 10
1263
BC117151.1
-
9, 10
1353
40125702
U78170.1
-
9, 10
1262
Y12336.1
-
9, 10
1355
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
signal transduction
Ras protein signal transduction
positive regulation of GTPase activity
cellular response to calcium ion
guanyl-nucleotide exchange factor activity
calcium ion binding
lipid binding
diacylglycerol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-462rt:80/10.239.167.23:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0