Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01000 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024368.1 | 5 | 1135 | |||
GenBank mRNA | AK302487.1 | - | 5 | 1156 | ||
BC062355.1 | - | 5 | 1054 | |||
BC131589.1 | - | 5 | 1054 |