Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286123.1 | NP_001273052.1 | 7 | 960 | ||
NM_001286125.1 | NP_001273054.1 | 7 | 960 | |||
NM_005835.3 | NP_005826.1 | 8 | 1410 | |||
XM_005248784.2 | XP_005248841.1 | 8 | 1410 | |||
XM_006714949.3 | XP_006715012.1 | 8 | 1147 | |||
XM_006714950.2 | XP_006715013.1 | 7 | 1037 | |||
XM_006714951.1 | XP_006715014.1 | 8 | 1410 | |||
XM_017010159.1 | XP_016865648.1 | 7 | 1300 | |||
XM_017010160.1 | XP_016865649.1 | 8 | 1405 | |||
GenBank mRNA | BC104822.1 | - | 7 | 933 | 8143825 | |
BC112033.1 | - | 7 | 933 | 8327501 | ||
BC143323.1 | - | 7 | 931 | |||
U90544.1 | - | 8 | 1295 |