Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RABL2A SIRNA Assays ›
Gene Symbol:
RABL2A
Gene Name:
RAB, member of RAS oncogene family-like 2A
Gene Aliases:
-
Chromosome Location:
Chr.2: 113627218 - 113643398 on Build GRCh38
UniGene ID:
Hs.446425
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s200490
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RABL2A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306158.1
NP_001293087.1
5, 6
530
NM_001306159.1
NP_001293088.1
5, 6
530
NM_001306160.1
NP_001293089.1
4, 5
358
NM_007082.4
NP_009013.1
5, 6
530
NM_013412.3
NP_038198.1
5, 6
530
XM_005263569.3
XP_005263626.1
5, 6
541
XM_005263570.3
XP_005263627.1
5, 6
508
XM_006712207.3
XP_006712270.1
5, 6
610
XM_006712208.3
XP_006712271.1
5, 6
499
XM_006712209.3
XP_006712272.1
5, 6
653
XM_006712215.3
XP_006712278.1
6, 7
740
XM_011510509.2
XP_011508811.1
5, 6
501
XM_017003203.1
XP_016858692.1
4, 5
1250
XM_017003204.1
XP_016858693.1
5, 6
1190
XM_017003205.1
XP_016858694.1
4, 5
1250
XM_017003206.1
XP_016858695.1
6, 7
651
XM_017003207.1
XP_016858696.1
5, 6
564
XM_017003208.1
XP_016858697.1
5, 6
524
XM_017003209.1
XP_016858698.1
5, 6
508
XM_017003210.1
XP_016858699.1
4, 5
1250
XM_017003211.1
XP_016858700.1
5, 6
541
XM_017003212.1
XP_016858701.1
4, 5
1250
XM_017003213.1
XP_016858702.1
4, 5
1250
XM_017003214.1
XP_016858703.1
5, 6
1190
XM_017003215.1
XP_016858704.1
4, 5
1250
XM_017003216.1
XP_016858705.1
5, 6
653
XM_017003217.1
XP_016858706.1
5, 6
537
XM_017003218.1
XP_016858707.1
6, 7
691
XM_017003219.1
XP_016858708.1
5, 6
604
XM_017003220.1
XP_016858709.1
4, 5
1250
XM_017003221.1
XP_016858710.1
5, 6
526
XM_017003222.1
XP_016858711.1
6, 7
681
XM_017003229.1
XP_016858718.1
5, 6
517
XM_017003230.1
XP_016858719.1
5, 6
531
XM_017003231.1
XP_016858720.1
5, 6
545
GenBank mRNA
AF095350.1
-
5, 6
458
AF095351.1
-
5, 6
458
AK289496.1
-
6, 7
445
BC126129.1
-
4, 5
372
8991845
BC144050.1
-
4, 5
372
9052570
BC144052.1
-
4, 5
372
9052572
BI552699.1
-
5, 6
537
CR456554.1
-
4, 5
365
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
GTPase activity
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0