Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CRLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012288.2 | NP_001012288.2 | 3 | 370 | ||
NM_022148.3 | NP_071431.2 | 4 | 473 | |||
NR_110830.1 | 4 | 470 | ||||
XM_011546181.2 | XP_011544483.1 | 4 | 508 | |||
XM_011546182.2 | XP_011544484.1 | 4 | 512 | |||
GenBank mRNA | AB052639.1 | - | 4 | 384 | ||
AF142570.1 | - | 4 | 400 | |||
AF338733.1 | - | 4 | 384 | |||
AK026800.1 | - | 4 | 472 | |||
AK223303.1 | - | 4 | 500 | |||
AK225583.1 | - | 4 | 472 | |||
AY775789.1 | - | 3 | 281 |