Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2AFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012412.4 | NP_036544.1 | 2 | 212 | ||
NM_138635.3 | NP_619541.1 | 2 | 212 | |||
NM_201516.2 | NP_958924.1 | 2 | 212 | |||
NM_201517.2 | NP_958925.1 | 2 | 212 | |||
GenBank mRNA | AB209001.1 | - | 2 | 123 | ||
AF081192.1 | - | 2 | 77 | |||
AK023973.1 | - | 2 | 163 | |||
AK129989.1 | - | 2 | 132 | |||
AL520811.3 | - | 2 | 113 | |||
BC000098.2 | - | 2 | 139 | 3505930 | ||
BC004274.1 | - | 1 | 64 | 3615320 | ||
BC014885.2 | - | 2 | 108 | 3907411 | ||
BC070169.1 | - | 2 | 139 | 6458566 | ||
BF242439.1 | - | 2 | 226 | |||
BG754026.1 | - | 2 | 187 | |||
BG773256.1 | - | 2 | 215 | |||
BI257891.1 | - | 2 | 170 | |||
BM452844.1 | - | 2 | 93 | |||
BM453663.1 | - | 2 | 82 | |||
CB148943.1 | - | 2 | 166 | |||
DB460745.1 | - | 2 | 212 |