Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOBEC3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145298.5 | NP_660341.2 | 5 | 880 | ||
XM_011529994.2 | XP_011528296.1 | 4 | 1151 | |||
GenBank mRNA | BC038808.1 | - | 5 | 665 | 5227163 | |
CX165430.1 | - | 5 | 671 | |||
JF262037.1 | - | 5 | 587 | |||
KC329530.1 | - | 5 | 587 |