Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282540.1 | NP_001269469.1 | 11 | 2143 | ||
NM_006382.3 | NP_006373.2 | 11 | 2143 | |||
GenBank mRNA | AK057407.1 | - | 10 | 1907 | ||
BC143291.1 | - | 4 | 419 | |||
U65652.1 | - | 4 | 672 |