Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148934.2 | NP_683736.1 | 2 | 431 | ||
GenBank mRNA | AF303106.1 | - | 2 | 431 | ||
AK153467.1 | - | 2 | 358 | |||
BC138515.1 | - | 2 | 472 | 8861535 | ||
BC138516.1 | - | 2 | 472 | 8861536 |