Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138649.1 | NP_619590.1 | 7 | 1306 | ||
GenBank mRNA | AB069667.1 | - | 7 | 1306 | ||
AK135098.1 | - | 5 | 1100 | |||
AK144560.1 | - | 7 | 1483 | |||
AK145293.1 | - | 7 | 1512 |