Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync2h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029851.2 | NP_084127.2 | 63 | 9836 | ||
GenBank mRNA | AK047928.1 | - | 18 | 2215 | ||
AK173324.1 | - | 5 | 507 | |||
AY452067.1 | - | 5 | 524 | |||
DQ104402.1 | - | 63 | 9836 |