Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005870.4 | NP_005861.2 | 2, 3 | 275 | ||
GenBank mRNA | AF153608.1 | - | 2, 3 | 213 | ||
AK311748.1 | - | 2, 3 | 196 | |||
AY550970.1 | - | 2, 3 | 195 | |||
BC030836.1 | - | 2, 3 | 243 | 4794888 | ||
CR457035.1 | - | 2, 3 | 179 | |||
DB171768.1 | - | 2, 3 | 277 | |||
U78303.1 | - | 2, 3 | 210 | |||
U96915.1 | - | 2, 3 | 195 |