Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007480.1 | NP_031506.1 | 3, 4 | 309 | ||
GenBank mRNA | BC132452.1 | - | 4, 5 | 297 | 40130729 | |
BC138756.1 | - | 4, 5 | 297 | 8861778 | ||
D87902.1 | - | 3, 4 | 309 |