Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007611.2 | NP_031637.1 | 3 | 406 | ||
GenBank mRNA | AK154978.1 | - | 3 | 285 | ||
BC005428.1 | - | 3 | 263 | 3600155 | ||
CT010348.1 | - | 2 | 217 | |||
D86353.1 | - | 3 | 269 | |||
Y13088.1 | - | 3 | 406 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | U67321.1 | - | 801 | Chr. - - [-] on Build |