Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007641.5 | NP_031667.1 | 2 | 182 | ||
GenBank mRNA | AK017903.1 | - | 2 | 183 | ||
AK041428.1 | - | 2 | 181 | |||
AK080040.1 | - | 2 | 148 | |||
AK080148.1 | - | 180 | ||||
AK137469.1 | - | 2 | 171 | |||
AK143824.1 | - | 2 | 177 | |||
AK156033.1 | - | 181 | ||||
AK166571.1 | - | 2 | 275 | |||
AK166659.1 | - | 2 | 275 | |||
AK171631.1 | - | 2 | 211 | |||
AK171638.1 | - | 2 | 134 | |||
AK171659.1 | - | 2 | 167 | |||
BC028322.1 | - | 2 | 163 | 4503841 | ||
CT010253.1 | - | 1 | 63 | |||
M62541.1 | - | 240 |