Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005877.5 | NP_005868.1 | 4 | 789 | ||
GenBank mRNA | AK303273.1 | - | 5 | 714 | ||
BC001976.1 | - | 4 | 729 | 3459964 | ||
BC007684.2 | - | 4 | 756 | 3634035 | ||
BI827130.1 | - | 4 | 548 | |||
CR456575.1 | - | 4 | 732 | |||
X85237.1 | - | 4 | 731 |