Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005877.5 | NP_005868.1 | 6 | 1014 | ||
GenBank mRNA | AK303273.1 | - | 7 | 939 | ||
BC001976.1 | - | 6 | 954 | 3459964 | ||
BC007684.2 | - | 6 | 981 | 3634035 | ||
CR456575.1 | - | 6 | 957 | |||
X85237.1 | - | 6 | 956 |