Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctla2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145801.1 | NP_001139273.1 | 3 | 334 | ||
NM_007797.1 | NP_031823.1 | 3 | 334 | |||
GenBank mRNA | AK150917.1 | - | 3 | 293 | ||
AY034578.1 | - | 3 | 274 | |||
BE448128.1 | - | 3 | 334 | |||
DT925153.1 | - | 23 | ||||
DT929456.1 | - | 24 | ||||
X15592.1 | - | 3 | 274 |
Gene Symbol: Ctla2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145799.1 | NP_001139271.1 | 3 | 318 | ||
NM_007796.2 | NP_031822.2 | 3 | 356 | |||
GenBank mRNA | AK005471.1 | - | 3 | 293 | ||
AK018801.1 | - | 3 | 311 | |||
AK132290.1 | - | 3 | 287 | |||
AK167433.1 | - | 3 | 289 | |||
AK167664.1 | - | 3 | 290 | |||
AY034577.1 | - | 3 | 265 | |||
BC028437.1 | - | 3 | 282 | 1244311 | ||
X15591.1 | - | 3 | 265 |