Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1b26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010644.2 | NP_034774.1 | 2 | 179 | ||
GenBank mRNA | BC024688.1 | - | 2 | 153 | 4167696 | |
BC138704.1 | - | 2 | 161 | 8861726 | ||
K01831.1 | - | 2 | 179 | |||
X63327.1 | - | 2 | 158 |
Gene Symbol: Egfbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010115.6 | NP_034245.3 | 2 | 161 | ||
GenBank mRNA | AB016032.1 | - | 2 | 137 | ||
X58628.1 | - | 2 | 161 |