Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008091.3 | NP_032117.1 | 2 | 733 | ||
GenBank mRNA | AK053728.1 | - | 1 | 1140 | ||
AK090089.1 | - | 2 | 741 | |||
AK155549.1 | - | 2 | 734 | |||
AK156634.1 | - | 2 | 720 | |||
AK157625.1 | - | 353 | ||||
AK157939.1 | - | 2 | 607 | |||
AK161099.1 | - | 2 | 611 | |||
BC062915.1 | - | 1 | 329 | 6826352 | ||
X55123.1 | - | 1 | 349 |