Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010272.1 | NP_034402.1 | 3 | 1174 | ||
GenBank mRNA | AF092734.1 | - | 3 | 919 | ||
KC561772.1 | - | 2286 |