Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010477.4 | NP_034607.3 | 6, 7 | 866 | ||
GenBank mRNA | AK088844.1 | - | 6, 7 | 780 | ||
AK143882.1 | - | 6, 7 | 771 | |||
AK146831.1 | - | 6, 7 | 818 | |||
AK177219.1 | - | 3, 4 | 291 | |||
AK179525.1 | - | 3, 4 | 290 | |||
AK187612.1 | - | 3, 4 | 296 | |||
AK188312.1 | - | 3, 4 | 289 | |||
AK216069.1 | - | 3, 4 | 260 | |||
AK217800.1 | - | 3, 4 | 288 | |||
BC016400.1 | - | 6, 7 | 752 | 3585270 | ||
BC106112.1 | - | 6, 7 | 767 | |||
X55023.1 | - | 6, 7 | 728 |