Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010478.2 | NP_034608.2 | 1 | 2694 | ||
GenBank mRNA | AK159139.1 | - | 2662 | |||
AK165042.1 | - | 1 | 2701 |