Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010646.1 | NP_034776.1 | 1 | 136 | ||
GenBank mRNA | AF204265.1 | - | 1 | 79 | ||
AF204266.1 | - | 1 | 79 | |||
AF204267.1 | - | 1 | 79 | |||
AF204268.1 | - | 1 | 79 | |||
AF307947.1 | - | 1 | 131 | |||
AF307948.1 | - | 1 | 136 |
Gene Symbol: Klra13-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF074460.1 | - | 1 | 84 | ||
AF283252.1 | - | 85 | ||||
BC099861.1 | - | 80 | 40040023 |
Gene Symbol: Klra23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024470.1 | NP_077790.1 | 2 | 202 | ||
GenBank mRNA | AF074459.1 | - | 2 | 202 | ||
AF074463.1 | - | 2 | 202 | |||
AF283250.1 | - | 1 | 85 | |||
AF283251.1 | - | 1 | 85 |