Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt33b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013570.1 | NP_038598.1 | 7 | 1349 | ||
GenBank mRNA | X75650.1 | - | 1063 |