Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap9-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015741.2 | NP_056556.1 | 1 | 531 | ||
GenBank mRNA | BC116210.2 | - | 1 | 480 | 40091112 |
Gene Symbol: Gm11567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101613.1 | NP_001095083.1 | 1 | 561 | ||
GenBank mRNA | AK003994.1 | - | 1 | 562 |