Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013825.3 | NP_038853.2 | 20, 21 | 2747 | ||
GenBank mRNA | AF395445.1 | - | 20, 21 | 2747 | ||
BC150734.1 | - | 2768 | 9087645 | |||
U19271.1 | - | 2781 |