Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265589.1 | NP_001252518.1 | 4, 5 | 2933 | ||
NM_001265590.1 | NP_001252519.1 | 3, 4 | 2597 | |||
NM_006054.3 | NP_006045.1 | 2, 3 | 545 | |||
NM_201428.2 | NP_958831.1 | 3, 4 | 2876 | |||
NM_201429.2 | NP_958832.1 | 3, 4 | 602 | |||
NM_201430.2 | NP_958833.1 | 2, 3 | 545 | |||
XM_011544730.2 | XP_011543032.1 | 4, 5 | 3023 | |||
XM_011544731.2 | XP_011543033.1 | 3, 4 | 737 | |||
GenBank mRNA | AB209771.1 | - | 2, 3 | 482 | ||
AF059524.1 | - | 2, 3 | 482 | |||
AF119297.1 | - | 2, 3 | 494 | |||
AK075412.1 | - | 2, 3 | 495 | |||
AK094965.1 | - | 2, 3 | 1598 | |||
AK122727.1 | - | 2, 3 | 998 | |||
AK127079.1 | - | 3, 4 | 2572 | |||
AK222898.1 | - | 2, 3 | 482 | |||
AK295361.1 | - | 3, 4 | 507 | |||
AK295534.1 | - | 2, 3 | 399 | |||
AK295758.1 | - | 3, 4 | 2525 | |||
AK304100.1 | - | 4, 5 | 2883 | |||
AK307811.1 | - | 3, 4 | 2822 | |||
AK308083.1 | - | 2, 3 | 490 | |||
AK309766.1 | - | 3, 4 | 2551 | |||
AY427821.2 | - | 3, 4 | 2843 | |||
AY750848.1 | - | 4, 5 | 2900 | |||
BC000634.1 | - | 2, 3 | 453 | |||
BC010556.1 | - | 2, 3 | 458 | |||
BC011394.1 | - | 2, 3 | 494 | |||
BC022993.1 | - | 2, 3 | 484 | |||
BC100822.2 | - | 2, 3 | 475 | |||
BC100823.2 | - | 2, 3 | 475 | |||
BC105981.1 | - | 2, 3 | 426 | |||
BC105982.1 | - | 2, 3 | 457 | |||
BC118550.1 | - | 2, 3 | 475 | |||
BC118628.1 | - | 2, 3 | 475 |