Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mafg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010756.3 | NP_034886.1 | 3 | 668 | ||
GenBank mRNA | AB221541.1 | - | 2 | 454 | ||
BC002092.1 | - | 3 | 606 | 3488374 | ||
BC052633.1 | - | 3 | 668 | 6309003 |